Order Entry
Canada
ContactUsLinkComponent
Masterflex® Adapter for 25, 50, and 100 mL Hamilton Gastight Syringe
Masterflex® Adapter for 25, 50, and 100 mL Hamilton Gastight Syringe
Catalog # MFLX74905-84
Supplier:  VWR International
undefined
Masterflex® Adapter for 25, 50, and 100 mL Hamilton Gastight Syringe
Catalog # MFLX74905-84
Supplier:  VWR International

About this item

Make the most of your Masterflex® syringe pumps with these accessories.

  • Optimize performance of your Masterflex pumps

Parts and accessories for Masterflex syringe pumps.

  • Description:
    Adapter for 25, 50, and 100 ml Hamilton Gastight Syringe
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Cat. no.:
    MFLX74905-84
Frequently Bought Together