Order Entry
Canada
ContactUsLinkComponent
 
Masterflex® Syringe Pump Line Cord, US Adapter; 115 VAC
  MFLX74905-78
undefined
Masterflex® Syringe Pump Line Cord, US Adapter; 115 VAC
  MFLX74905-78

 

Make the most of your Masterflex® syringe pumps with these accessories.

  • Optimize performance of your Masterflex pumps

Parts and accessories for Masterflex syringe pumps.

  • Description:
    Syringe Pump Line Cord (US - 115V)
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Cat. no.:
    MFLX74905-78
Frequently Bought Together