ContactUsLinkComponent
Masterflex® Mechanically Coupled Centrifugal Pump Flange Kit for 1 1/2" and 2" Connections
Masterflex® Mechanically Coupled Centrifugal Pump Flange Kit for 1 1/2" and 2" Connections
Catalog # MFLX70725-72
Supplier:  VWR International
undefined
Masterflex® Mechanically Coupled Centrifugal Pump Flange Kit for 1 1/2" and 2" Connections
Catalog # MFLX70725-72
Supplier:  VWR International
  • Wetted materials:
  • SS flanges, gaskets, nuts, bolts, flat washer, lockwashers
  • SS flanges, gaskets, nuts, bolts, flat washer, lockwashers
  • Warranty:
  • 1 year
  • 1 year
  • Currently Viewing
Specifications
About this item
Enhance the life of your pump with service flange kits.
  • Constructed of stainless steel (SS) parts
  • Fits 1 1/2" and 2" connections
Flange kits are competible with your 304 SS mechanically coupled centrifugal pumps.
Delivery: Includes stainless steel 150# threaded RF companion flanges, gaskets, nuts, bolts, flat washer, and lock washers.
  • Description:
    Flange Kit for 1-1/2" and 2" Connections
  • Environmentally Preferable:
  • Wetted materials:
    SS flanges, gaskets, nuts, bolts, flat washer, lockwashers
  • Warranty:
    1 year
  • Cat. no.:
    MFLX70725-72
Frequently Bought Together