Masterflex® Plastic Assorted Fitting Kits, Avantor®
Fitting Kit, PVDF, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 Set, Kit, Straight, PVDF
Masterflex® Plastic Assorted Fitting Kits, Avantor®
Catalog # MFLX31514-36
CE certificate
Jump to:
Product Details & Documents
Get organized today!
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Assorted fittings come in an impact-resistant plastic box with secure snap-shut latches to prevent spillage
- Labeled compartments let you easily locate the fittings you want
- Order storage racks separately
Organize your workstation with these convenient fitting kits. Assorted fittings come in impact resistant plastic box with secure latch to keep fittings organized and in place. Label compartments make fitting location quick and easy. Choose from four materials: high-density polyethylene (HDPE), black nylon, natural polypropylene, and natural PVDF.
Documents
Certificates
Search by Lot NumberSpecifications
- Connection Type:Kit
- Flow pattern:Straight
- Description:Fitting Kit, PVDF, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 Set
- Fitting Material:PVDF
- Cat. No.:MFLX31514-36
- Warranty:3 Months
- Min. temperature:–62 °C (–79 °F)
- Max. temperature:135 °C (275 °F)
- Model:31514-36
Accessories
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, Black Nylon, Straight and Reducer, Hose Barb, Large Fittings; 1 SetFitting Kit, Black Nylon, Straight and Reducer, Hose Barb, Large Fittings; 1 SetMFLX31514-06KitStraightLog in for availabilityEach - $177.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, Black Nylon, Straight and Reducer, Thread Adapter to HosebarbFitting Kit, Black Nylon, Straight and Reducer, Thread Adapter to HosebarbMFLX31514-10KitStraightLog in for availabilityEach - $205.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, Black Nylon, Elbow/Tee/Y, Hose Barb; 1 SetFitting Kit, Black Nylon, Elbow/Tee/Y, Hose Barb; 1 SetMFLX31514-14KitElbowLog in for availabilityEach - $198.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, Black Nylon, Elbow and Tee, Hose Barb; 1 SetFitting Kit, Black Nylon, Elbow and Tee, Hose Barb; 1 SetMFLX31514-18KitTLog in for availabilityEach - $168.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, Black Nylon, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetFitting Kit, Black Nylon, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetMFLX31514-26KitTLog in for availabilityEach - $206.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, HDPE, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetFitting Kit, HDPE, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetMFLX31514-25KitTLog in for availabilityEach - $243.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, HDPE, Straight/Reducer/Elbow/Tee, BSP Threaded Adapter to HosebarbFitting Kit, HDPE, Straight/Reducer/Elbow/Tee, BSP Threaded Adapter to HosebarbMFLX31514-32KitTLog in for availabilityEach - $1,090.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, Black Nylon, Straight/Reducer/Elbow/Tee, BSP Thread to HosebarbFitting Kit, Black Nylon, Straight/Reducer/Elbow/Tee, BSP Thread to HosebarbMFLX31514-29KitTLog in for availabilityEach - $205.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, HDPE, Straight and Reducer, Threaded Adapter to Hose Barb; 1 SetFitting Kit, HDPE, Straight and Reducer, Threaded Adapter to Hose Barb; 1 SetMFLX31514-09KitStraightLog in for availabilityEach - $206.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, HDPE, Straight and Reducer, Hose Barb, Large FittingsFitting Kit, HDPE, Straight and Reducer, Hose Barb, Large FittingsMFLX31514-05KitStraightLog in for availabilityEach - $184.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, HDPE, Elbow/Tee/Y, Hose Barb; 1 SetFitting Kit, HDPE, Elbow/Tee/Y, Hose Barb; 1 SetMFLX31514-13KitElbowLog in for availabilityEach - $247.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, HDPE, Elbow and Tee, Hose Barb; 1 SetFitting Kit, HDPE, Elbow and Tee, Hose Barb; 1 SetMFLX31514-17KitTLog in for availabilityEach - $219.00
Product Information
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Fitting Kit, Black Nylon, Elbow/Tee/Nipple/Plug, Thread Adapter to HosebarbFitting Kit, Black Nylon, Elbow/Tee/Nipple/Plug, Thread Adapter to HosebarbMFLX31514-22KitElbowLog in for availabilityEach - $182.00