BASIC* Lab Coats, Halyard
White, XXL
CA76540-360
: 10124
HALYARD* BASIC* Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
- Traditional collar
- Raglan sleeves
- Knit cuffs
- Four snap front closure
- Two reinforced pockets
- Size/name label
- Clothing Size:XXL
- Color:White