Orders LinkContactUsLinkComponent
High Capacity Racking Systems
High Capacity Racking Systems
Stainless steel 17-rack system for 31" diameter freezer
Catalog # 76542-394
Supplier #:  3101S-C100
High Capacity Racking Systems
Catalog # 76542-394
High Capacity Racking Systems
High Capacity Racking Systems
Stainless steel 17-rack system for 31" diameter freezer
Price
3 724,17 $
Log in for availability
Each
Product Details & Documents
Our racking systems are precision-engineered to provide maximum storage capacity, organization, and durability inside laboratory and isothermal freezers.
  • Choice of materials
  • Stainless steel: Provides long-term corrosion resistance, strength, and durability under repeated freeze-thaw cycles. Ideal for high-demand research or GMP environments
  • Aluminum: Lightweight yet robust for easy handling and installation, offering excellent thermal conductivity and reduced mass for fast temperature recovery
  • Lifetime warranty on workmanship
Designed to fit a variety of freezer diameters - including 20, 21, 31 and 39-inch isothermal freezer models - these systems are ideal for laboratories, biobanks, and research facilities requiring efficient cryogenic sample management.
Constructed from stainless steel or aluminum, each system delivers superior corrosion resistance and structural integrity under ultra-low and cryogenic temperature conditions. Whether used in vapor-phase liquid nitrogen (LN₂) storage or mechanical freezer environments, these racks maintain consistent performance and easy accessibility during long-term sample storage.
Specifications
  • Description:
    Stainless steel 17-rack system for 31" diameter freezer