Orders LinkContactUsLinkComponent
Sequential Sterilization Wraps, Halyard
H200 sterilization wrap, 40×40"
Catalog # CA76540-232
 :  68040
Sequential Sterilization Wraps, Halyard
Catalog # CA76540-232
Sequential Sterilization Wraps, Halyard
Sequential Sterilization Wraps, Halyard
H200 sterilization wrap, 40×40"
Price
$397.87
Log in for availability
Case of 250
Product Details & Documents
Wrap made of patented low-lint SMS sterilization fabric that is tear-, flame-, lint-, and abrasion-resistant. Recommended for light linen packs, linen gowns, towel packs, single basins, and other light wrapping.
  • Industry standard for sterile processing teams around the world
  • POWERGUARD* technology for enhanced microbial barrier protection
  • Pre-vacuum steam sterilant penetration
  • Ethylene Oxide (EO) sterilant penetration and residuals
  • Class 1 flammability rating
Since 1978, the HALYARD* Sterilization Wrap, made of strong reliable SMS nonwoven fabric, has been helping keep sterilized instruments and supply sets ready for use at a moment’s notice. The company pioneered the development of SMS fabric and today is one of the largest global suppliers. It’s good to know that individual medical and dental practices can also take advantage of the same performance demanded by major medical and surgery centers. All HALYARD* products are made in the USA.
Today, Halyard Health offers six grades of wrap fabric, from H100 to H500, to offer the precise protection required for specific needs. Choosing is easy, the higher the number, the stronger the wrap. HALYARD* is also available in smaller sizes that meet the needs of smaller dental or medical offices.
 : Maintenance of package integrity (MPI) to meet FDA requirements. Instructions for Use to meet FDA guidelines.
Specifications
  • Description:
    H200 sterilization wrap
  • Dimensions:
    40×40"