CAPSULE FILTER MINI 25CSPSS0022S PK4
Catalog # 76541-138
Supplier: PARKER HANNIFIN TE
CAPSULE FILTER MINI 25CSPSS0022S PK4
Catalog # 76541-138
Supplier: PARKER HANNIFIN TE
Supplier Number: 25-CSPSS-002-2-S
Frequently Bought Together
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Leakbuster™ Specimen Containers
Supplier: Starplex
Betaine enhancer solution 5M
Supplier: VWR International
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
Water, deionized VWR®
Supplier: VWR
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Water for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Sterile 70% Isopropanol
Supplier: VWR International
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Phenolphthalein 1% (w/v) in 95% (v/v) alcohol indicator, neutralized
Supplier: Ricca Chemical
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics
Corning® Cell Recovery Solution, Corning
Supplier: Corning
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning