Styrofoam Head Models
Faceless, 5.8×6.8×9.8"
Catalog # 470337-668
Supplier: FLORA CRAFT CORPORATION
Supplier #: RS271/6
Jump to:
Product Details & Documents
Styrofoam head models for multiple uses.
- Available in male, female, and faceless versions
- Smooth surface paints easily
- Dense and durable
These smooth foam head forms can be painted or finished, allowing students to make or display their own models. Use them as a base to create brain maps, or sculpt masks. The foam is denser than most styrofoam, so it can also be trimmed or cut to create internal anatomical models. They may also be used for basic modeling projects in multiple disciplines.
Specifications
- Description:Faceless
- Size:5.8×6.8×9.8"
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.