Manual Pinch Valve, 5/32" to 1/4" tube OD
Catalog # MFLX98002-00
Supplier: VWR International
Specifications
See all product configurations- Catalog No:
- MFLX98002-00
- MFLX98002-20
- MFLX98002-10
- Pinch valve tube size:
- 5/32" to 1/4" OD
- 7/16" to 1/2" OD
- 5/16" to 3/8" OD
- Warranty:
- 3 Months
- 3 Months
- 3 Months
- Currently Viewing
Specifications
About this item
An ideal option to secure tubing.
- Constructed of durable Delrin®
Fluid contacts only the tubing—ideal for use in pharmaceutical and food industries where eliminating contamination is critical. Obtain precise and repeatable flow rates using easy-to-read metered scale on valve body. Valves are made of Delrin®. Tubing is not included—order flexible tubing with durometer (Shore A) of 60 or less from our "Tubing" section.
- Connection Type:Pinch Valve for Tubing
- Description:Manual Pinch Valve, 5/32" to 1/4" tube OD
- Environmentally Preferable:
- Pinch valve tube size:5/32" to 1/4" OD
- Warranty:3 Months
- Cat. no.:MFLX98002-00
Frequently Bought Together