Order Entry
Canada
ContactUsLinkComponent
Masterflex® Ismatec® Footswitch for BVP and MCP Series Drives, DB-15 Male Connector; 6-ft (1.8 m)
Masterflex® Ismatec® Footswitch for BVP and MCP Series Drives, DB-15 Male Connector; 6-ft (1.8 m)
Catalog # MFLX78003-07
undefined
Masterflex® Ismatec® Footswitch for BVP and MCP Series Drives, DB-15 Male Connector; 6-ft (1.8 m)
Catalog # MFLX78003-07
Supplier Number:  IS10039A

About this item

Simplify your dispensing.

  • Select the footswitch with connectio suitable for your Ismatec® pump

Footswitches provide convenient, hands-free operation of your pump drives from up to six feet away. Simply press once to start pump, press again to stop.

  • Description:
    Footswitch for BVP and MCP Series Drives, DB-15 Male Connector; 6-ft (1.8 m)
  • Length:
    183 cm (72")
  • Environmentally Preferable:
  • Model:
    78003-07
  • Input:
    DB-15
  • Warranty:
    1 year
  • Series:
    Ismatec BVP and MCP Drives
  • Cat. no.:
    MFLX78003-07
Frequently Bought Together