ContactUsLinkComponent
Masterflex® I/P® Replacement Main Body, PPS Housing, CRS Rotor
Masterflex® I/P® Replacement Main Body, PPS Housing, CRS Rotor
Catalog # MFLX77602-07
undefined
Masterflex® I/P® Replacement Main Body, PPS Housing, CRS Rotor
Catalog # MFLX77602-07
Supplier Number:  77602-07
About this item
Get the most our of your Masterflex® I/P® Easy-Load® Pump Head with these replacement parts.
  • Suitable for use with pump heads 77601-60 and 77601-62
Keep your pump head operating at its best with these replacement main body and occusion bed assemblies.
  • Pump series:
    I/P
  • Description:
    Replacement Main Body, PPS Housing, CRS Rotor
  • Environmentally Preferable:
  • Warranty:
    3 Months
  • Cat. no.:
    MFLX77602-07
  • Rotor Material:
    Cold-rolled steel
  • Housing Material:
    PPS
Frequently Bought Together