Masterflex® L/S® Power Supply, Dual Voltage 115/230 VAC to 12 VDC, 12-ft Cable
Catalog # MFLX77200-07
Supplier: Avantor Fluid Handling
About this item
Make the most of your Masterflex drives replacement parts and other accessories.
- Suitable for use with 07571-00 L/S drive
This power supply is rated for 3.0 Amps at 12 VDC. The total output power is 36 watts at 12 VDC.
- Description:Power supply, dual voltage 115/230 VAC to 12 VDC, 12-ft cable
- Environmentally Preferable:
- Warranty:1 year
- Compatible with:07571-00 L/S drive
- Cat. no.:MFLX77200-07
Frequently Bought Together