Masterflex® Replacement RJ-12 Cable for Liqui-Sense Sensor, 6-pin; 3.5 ft
Catalog # MFLX77095-02
Supplier: Avantor Fluid Handling
About this item
Make the most of your Liqui-Sense leak detector with suitable replacement parts.
- Compatible with Masterflex® Liqui-Sense controllers
This replacement cable features a 6-pin cable with RJ-12 connections and has a 3.5 foot cable length.
- Description:Replacement RJ-12 cable
- Length:3.5 ft.
- Environmentally Preferable:
- Model:77095-02
- Cat. no.:MFLX77095-02
Frequently Bought Together