Masterflex® Communication Cable, Pump to Pump, RS-485, 6 ft (2 m)
Catalog # MFLX74905-74
Supplier: VWR International
About this item
Make the most of your Masterflex® syringe pumps with these accessories.
- Optimize performance of your Masterflex pumps
Parts and accessories for Masterflex syringe pumps.
- Description:Communication Cable, pump to pump, RS-485, 6 ft (2 m)
- Environmentally Preferable:
- Warranty:1 year
- Cat. no.:MFLX74905-74
Frequently Bought Together