Order Entry
Canada
ContactUsLinkComponent
Masterflex® Communication Cable, Pump to Pump, RS-485, 1.6 ft (0.5 m)
Masterflex® Communication Cable, Pump to Pump, RS-485, 1.6 ft (0.5 m)
Catalog # MFLX74905-73
Supplier:  VWR International
undefined
Masterflex® Communication Cable, Pump to Pump, RS-485, 1.6 ft (0.5 m)
Catalog # MFLX74905-73
Supplier:  VWR International

About this item

Make the most of your Masterflex® syringe pumps with these accessories.

  • Optimize performance of your Masterflex pumps

Parts and accessories for Masterflex syringe pumps.

  • Description:
    Communication Cable, pump to pump, RS-485, 1,6 ft (0,5 m)
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Cat. no.:
    MFLX74905-73
Frequently Bought Together