Masterflex® Touch-Screen Continuous-Cycle Four-Syringe Pump Accessories, Avantor®
Syringe Pump Continuous Medium Pressure PTFE Pinch Valve 75 psi
Catalog # MFLX74901-93
CE Compliant
Masterflex® Touch-Screen Continuous-Cycle Four-Syringe Pump Accessories, Avantor®
Catalog # MFLX74901-93
CE Compliant
Jump to:
Product Details & Documents
Make the most of your syringe pump with these accessories.
- Choose from a wide range of replacement parts and accessories
Optimize performance of your Masterflex® Touch-Screen, Continuous-Cycle, Four-Syringe Pump
Specifications
- Description:Syringe Pump Continuous Medium Pressure PTFE Pinch Valve 75 psi
- Cat. No.:MFLX74901-93
- Warranty:1 year
Accessories
Product Information
- Catalog #AvailabilityPrice
- Pumps, Syringe Pumps Daisy Chain Cable for 74901-80 Dual Drive Syringe PumpPumps, Syringe Pumps Daisy Chain Cable for 74901-80 Dual Drive Syringe PumpMFLX74901-87Log in for availabilityEach - $130.00
Product Information
- Catalog #AvailabilityPrice
- Pumps, Syringe Pumps RS-232 Connector Cable for 74901-80 Dual Drive Syringe PumpPumps, Syringe Pumps RS-232 Connector Cable for 74901-80 Dual Drive Syringe PumpMFLX74901-88Log in for availabilityEach - $321.00