ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Y-Connector, Hose Barb Union, ¹/₁₆" Int.Ø; 100 Pack
  MFLX40727-41
CE certificate
undefined
Masterflex® Fitting, Polypropylene, Y-Connector, Hose Barb Union, ¹/₁₆" Int.Ø; 100 Pack
  MFLX40727-41
CE certificate
  • Model:
  • 40727-41
  • 40726-49
  • 40726-50
  • 40727-47
  • 40727-45
  • 40727-46
  • 40727-43
  • 40727-44
  • 30614-71
  • 40727-42
  • 30614-67
  • 41526-39
  • 30614-63
  • 41526-28
  • 30614-51
  • 30614-59
  • 40726-48
  • 40726-46
  • 40726-47
  • 30614-43
  • 40726-44
  • 40726-45
  • 40726-42
  • 40726-43
  • 40726-41
  • Sterilisation:
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Gamma Irradiation, Autoclave, and EtO
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 1 year
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • Shelf life
  • Shelf life
  • Shelf life
  • Shelf life
  • Shelf life
  •  
No tools needed for assembly.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: 1/16" ID; 100/PK
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
  • Barb size/tubing int.Ø (port 1):
    ¹⁄₁₆"
  • Barb size/tubing int.Ø (port 2):
    ¹⁄₁₆"
  • Connection Type:
    Barbed Union
  • Flow pattern:
    Y
  • Description:
    Fitting, polypropylene, Y-connector, hose barb union, ¹/₁₆" Int.Ø; 100 pack
  • Environmentally Preferable:
  • Model:
    40727-41
  • Sterilisation:
    Gamma Irradiation, Autoclave, and EtO
  • Warranty:
    3 Months
  • Cat. no.:
    MFLX40727-41
  • Fitting Material:
    PP
  • Barb Size / Tubing I.D. (Port 3):
    ¹⁄₁₆"
Frequently Bought Together