Masterflex® Fitting, HDPE, Straight, Hose Barb Union, 1/8" ID; 100/PK
MFLX40626-54
Quick and easy assembly without tools.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory Approvals to match your unique application
- Ultra smooth inner surface results in fewer extractables
- Connection Type: 1/8" ID
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Barb size/tubing int.Ø (port 1):⅛"
- Barb size/tubing int.Ø (port 2):⅛"
- Connection Type:Barbed Union
- Flow pattern:Straight
- Description:Fitting, HDPE, Straight, Hose Barb Union, ⅛" ID; 100/PK
- Environmentally Preferable:
- Sterilisation:Ethylene oxide (ETO) only
- Cat. no.:MFLX40626-54
- Fitting Material:HDPE
Frequently Bought Together