Order Entry
Canada
ContactUsLinkComponent
McMillan Flow High-Accuracy Flowmeter Power Supply, 120 VAC 60 Hz to 12 VDC
McMillan Flow High-Accuracy Flowmeter Power Supply, 120 VAC 60 Hz to 12 VDC
Catalog # MFLX32704-55
Supplier:  MC MILLAN CO
CE Compliant
McMillan Flow High-Accuracy Flowmeter Power Supply, 120 VAC 60 Hz to 12 VDC
Catalog # MFLX32704-55
Supplier:  MC MILLAN CO
Supplier Number:  110-00-08T
CE Compliant

About this item

Ideal for use with McMillan Flow High-Accuracy Flowmeters.

  • A wide range of output options for integration into any system

These power supplies are required to power the Modular Liquid Flow Rate Sensor Systems.

  • Description:
    High-Accuracy Flowmeter Power Supply, 120 VAC 60 Hz to 12 VDC
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Cat. no.:
    MFLX32704-55
  • Power:
    120 VAC; 12 VDC
Frequently Bought Together