ContactUsLinkComponent
Parker Hannifin Compression Union Tee Adapter, SS, 1/4" OD (2)×1/4" NPT(M); Each
Parker Hannifin Compression Union Tee Adapter, SS, 1/4" OD (2)×1/4" NPT(M); Each
Catalog # MFLX31406-86
CE certificate
undefined
Parker Hannifin Compression Union Tee Adapter, SS, 1/4" OD (2)×1/4" NPT(M); Each
Catalog # MFLX31406-86
Supplier Number:  ISSD4MBT4
CE certificate
About this item
Get a secure seal at higher pressures.
  • Quick and easy assembly without tools
Compression and threaded fittings offer a more permanent installation. Choose the size and material to suit your application needs.
  • Connection Type:
    Compression to Thread
  • Flow pattern:
    T
  • Description:
    Compression Union Tee Adapter, SS, 1/4" OD (2)×1/4" NPT(M); Each
  • Environmentally Preferable:
  • Sterilisation:
    Autoclave, ethylene oxide (ETO), or gamma
  • Warranty:
    LTD life time
  • Cat. no.:
    MFLX31406-86
  • Fitting Material:
    Stainless steel 316
Frequently Bought Together