Parker Hannifin Compression Union Tee Adapter, SS, 1/8" OD (2)×1/8" NPT(M); Each
Catalog # MFLX31406-85
Supplier: MIDWEST VALVE & FITTING INC
CE certificate
About this item
Get a secure seal at higher pressures.
- Quick and easy assembly without tools
Compression and threaded fittings offer a more permanent installation. Choose the size and material to suit your application needs.
- Connection Type:Compression to Thread
- Flow pattern:T
- Description:Compression Union Tee Adapter, SS, 1/8" OD (2)×1/8" NPT(M); Each
- Environmentally Preferable:
- Sterilisation:Autoclave, ethylene oxide (ETO), or gamma
- Warranty:LTD life time
- Cat. no.:MFLX31406-85
- Fitting Material:Stainless steel 316
Frequently Bought Together