Masterflex® Fitting, Stainless Steel, Y-Connector, Hose Barb Union, 5/32" ID
Catalog # MFLX30620-21
Supplier: VWR International
Specifications
See all product configurations- Model:
- 30620-21
- 30620-23
- 31209-56
- 30620-22
- 31209-55
- 30620-24
- 31209-57
- Orifice dimensions:
- 0.03125"
- 0.0625"
- 0.28125"
- Sterilisation:
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Warranty:
- 1 Month
- 1 Month
- 1 year
- 1 Month
- 1 year
- 1 Month
- 1 year
- Currently Viewing
Specifications
About this item
Ensure a tight seal every time.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
- Connection Type: 5/32" ID; 1/EA
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
- Barb size/tubing int.Ø (port 1):⁵⁄₃₂"
- Barb size/tubing int.Ø (port 2):⁵⁄₃₂"
- Connection Type:Barbed Union
- Flow pattern:Y
- Description:Fitting, Stainless Steel, Y-Connector, Hose Barb Union, 5/32" ID
- Environmentally Preferable:
- Model:30620-21
- Sterilisation:Autoclave, ethylene oxide (ETO), or gamma
- Warranty:1 Month
- Cat. no.:MFLX30620-21
- Fitting Material:Stainless steel 316
- Barb Size / Tubing I.D. (Port 3):⁵⁄₃₂"
Frequently Bought Together