ContactUsLinkComponent
Masterflex® Fitting, Nylon, Straight, Hose Barb Reducers, 1/2" ID x 3/8" ID; 10/PK
Masterflex® Fitting, Nylon, Straight, Hose Barb Reducers, 1/2" ID x 3/8" ID; 10/PK
Catalog # MFLX30616-22
Supplier:  VWR International
undefined
Masterflex® Fitting, Nylon, Straight, Hose Barb Reducers, 1/2" ID x 3/8" ID; 10/PK
Catalog # MFLX30616-22
Supplier:  VWR International
About this item
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: ¹/₂" int.Ø × ³/₈" int.Ø
Hose barb fittings make tubing connections quick and easy, requiring no tools for installation. Select the fitting based on the Masterflex tubing size. Fittings are available in a variety of materials to suit different applications and budgets.
  • Barb size/tubing int.Ø (port 1):
    ½"
  • Barb size/tubing int.Ø (port 2):
    ⅜"
  • Connection Type:
    Barbed Reducer
  • Flow pattern:
    Straight
  • Description:
    Fitting, Nylon, Straight, Hose Barb Reducers, 1/2" ID x 3/8" ID; 10/PK
  • Environmentally Preferable:
  • Model:
    30616-22
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Cat. no.:
    MFLX30616-22
  • Fitting Material:
    Nylon
Frequently Bought Together