Order Entry
Canada
Orders LinkContactUsLinkComponent
PendoTECH® Single-Use Pressure Sensor Accessories

PendoTECH® Single-Use Pressure Sensor Accessories

RS232 Sensor Cable with 4-wire leads, 12 ft (3.7 m)
Catalog # MFLX19406-82
Supplier:   PENDOTECH
Supplier #:  PDKTP-RS232
PendoTECH® Single-Use Pressure Sensor Accessories
Catalog # MFLX19406-82
Supplier:  PENDOTECH
PendoTECH® Single-Use Pressure Sensor Accessories
PendoTECH® Single-Use Pressure Sensor Accessories

RS232 Sensor Cable with 4-wire leads, 12 ft (3.7 m)

Price
$163.00
Log in for availability
Each
Product Details & Documents

Optimize performance of your PendoTECH® Pressure Sensor.

  • Choose from a wide range of replacement parts and accessories

Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.

Specifications
  • Description:
    RS232 Sensor Cable with 4-wire leads, 12 ft (3.7 m)
  • Cat. No.:
    MFLX19406-82
  • Warranty:
    1 year
  • Length:
    365.76 cm (144")
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.