Order Entry
Canada
Orders LinkContactUsLinkComponent
 

PendoTECH® Single-Use Pressure Sensor Accessories

Sensor Cable with 4-wire leads, for 3rd party monitors 12ft (3,7 m)
Catalog # MFLX19406-80
Supplier:   PENDOTECH
 :  PDKT-650-298
PendoTECH® Single-Use Pressure Sensor Accessories
Catalog # MFLX19406-80
Supplier:  PENDOTECH
PendoTECH® Single-Use Pressure Sensor Accessories
PendoTECH® Single-Use Pressure Sensor Accessories

Sensor Cable with 4-wire leads, for 3rd party monitors 12ft (3,7 m)

Price
$185.00
Log in for availability
Each
Product Details & Documents

Optimize performance of your PendoTECH® Pressure Sensor.

  • Choose from a wide range of replacement parts and accessories

Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.

Specifications
  • Description:
    Sensor Cable with 4-wire leads, for 3rd party monitors 12ft (3,7 m)
  • Cat. No.:
    MFLX19406-80
  • Warranty:
    1 year
  • Length:
    365.76 cm (144")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.