ContactUsLinkComponent
Masterflex® L/S® Economy Variable-Speed Drive Replacement Brushes; 2/PK
  MFLX07520-06
undefined
Masterflex® L/S® Economy Variable-Speed Drive Replacement Brushes; 2/PK
  MFLX07520-06
 :  7520-06
Quickly and easily replace components of your Masterflex system.
  • Choose replacement brushes and caps based on your Masterflex L/S drive
  • Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.
  • Description:
    Replacement brushes for L/S drive series: 07554-80, -85, -90, -95
  • Environmentally Preferable:
  • Cat. no.:
    MFLX07520-06
Frequently Bought Together