ContactUsLinkComponent
Masterflex® L/S® Peristaltic Pump Replacement Brush Cap; 2/Set
Masterflex® L/S® Peristaltic Pump Replacement Brush Cap; 2/Set
Catalog # MFLX07520-03
Supplier:  Avantor
undefined
Masterflex® L/S® Peristaltic Pump Replacement Brush Cap; 2/Set
Catalog # MFLX07520-03
Supplier:  Avantor
About this item
Quickly and easily replace components of your Masterflex system.
  • Choose replacement brushes and caps based on your Masterflex L/S drive
  • Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.
Brush cap holder works with all 1/20 hp (37 W), 1/10 hp (75 W), and 1/5 hp (150 W) drive series.
  • Description:
    Drive Replacement Brush Cap
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Compatible with:
    Series 07520, 07521, 07522, 07523, 07525, 07526, 07527, 07528, 07553 (except -50, -60), 07554 (except -50, -60), 07558, 07567, 07568, 07592, 77300
  • Cat. no.:
    MFLX07520-03
Frequently Bought Together