ContactUsLinkComponent
Masterflex® I/P® Tubing Loading Key for Standard Pump Heads
Masterflex® I/P® Tubing Loading Key for Standard Pump Heads
Catalog # MFLX07019-90
undefined
Masterflex® I/P® Tubing Loading Key for Standard Pump Heads
Catalog # MFLX07019-90
Supplier Number:  7019-90
About this item
Ensure proper tubing alignment and installation.
  • Constructed of durable stainless steel
  • Tubing key is suitable for use with I/P® standard pump heads
L/S® and I/P® Standard pump heads include tubing loading keys. A key is required to ensure proper tubing alignment and tension. Order a replacement key based on your pump head series, L/S® or I/P®.
  • Description:
    tubing loading key for I/P Standard pump heads.
  • Environmentally Preferable:
  • Warranty:
    2 years
  • Cat. no.:
    MFLX07019-90
  • Pump Series:
    I/P
Frequently Bought Together