ContactUsLinkComponent
Masterflex® Nylon Clamp for Sanitary Fittings, 1-1-1/2" Clamp Size; 10/PK
Masterflex® Nylon Clamp for Sanitary Fittings, 1-1-1/2" Clamp Size; 10/PK
Catalog # MFLX00424-TO
Supplier:  VWR International
undefined
Masterflex® Nylon Clamp for Sanitary Fittings, 1-1-1/2" Clamp Size; 10/PK
Catalog # MFLX00424-TO
Supplier:  VWR International
About this item
Collared clamp helps prevent unintentional opening.
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
  • Connection type:
    Sanitary
  • Description:
    Nylon Clamp for Sanitary Fittings, 1-1-1/2" Clamp Size; 10/PK
  • Material:
    Nylon
  • Environmentally Preferable:
  • Sanitary clamp size (port 1):
    1-1/2"
  • Sterilisation:
    Autoclave, Gamma
  • Warranty:
    1 year
  • Cat. no.:
    MFLX00424-TO
  • Clamp Type:
    Sanitary
Frequently Bought Together