Orders LinkContactUsLinkComponent
Test Tubes without Rim
Test Tubes without Rim
175 ml, 38×200 mm
Catalog # 76532-236
Supplier #:  TT9820-L
Test Tubes without Rim
Catalog # 76532-236
Test Tubes without Rim
Test Tubes without Rim
175 ml, 38×200 mm
Price
$229.91
New Customer Promotional Price
Log in for availability
Pack of 50
Product Details & Documents
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
  • Tubes do not have rims
  • Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Specifications
  • Capacity:
    175 ml
  • Diameter ×H:
    38×200 mm