Orders LinkContactUsLinkComponent
Test Tubes without Rim
Test Tubes without Rim
14 ml, 16×100 mm
Catalog # 76532-226
Supplier #:  TT9820-F
Test Tubes without Rim
Catalog # 76532-226
Test Tubes without Rim
Test Tubes without Rim
14 ml, 16×100 mm
Price
$67.87
New Customer Promotional Price
Log in for availability
Pack of 72
Product Details & Documents
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
  • Tubes do not have rims
  • Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Specifications
  • Capacity:
    14 ml
  • Diameter ×H:
    16×100 mm