Orders LinkContactUsLinkComponent
Test Tubes without Rim
Test Tubes without Rim
17 ml, 15×150 mm
Catalog # 76532-224
Supplier #:  TT9820-E
Test Tubes without Rim
Catalog # 76532-224
Test Tubes without Rim
Test Tubes without Rim
17 ml, 15×150 mm
Price
$72.48
New Customer Promotional Price
Log in for availability
Pack of 72
Product Details & Documents
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
  • Tubes do not have rims
  • Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Specifications
  • Capacity:
    17 ml
  • Diameter ×H:
    15×150 mm