Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Vikan® Hygienic Wall Bracket, Single Hook Module, Remco

Black, 1.63×3.07×1.89"
Catalog # 76528-646
Supplier:   Remco Products
 :  10129
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Catalog # 76528-646
Supplier:  Remco Products
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco

Black, 1.63×3.07×1.89"

Price
$21.66
Log in for availability
Each
Product Details & Documents

Single hook module is designed for storing cleaning tools that have a hanging hole.

  • Can hold products weighing up to 6.61 lbs.
  • Polypropylene/polyamide construction

The hook system is easy to disassemble for cleaning or replacement.

Specifications
  • Color:
    Black
  • Dimensions:
    1.63×3.07×1.89"
  • Cat. No.:
    76528-646
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.