KIT REAGENT NINHYDRIN PK2
Catalog # 76522-128
Supplier: HITACHI HIGH-TECH AMERICA, INC.
KIT REAGENT NINHYDRIN PK2
Catalog # 76522-128
Supplier: HITACHI HIGH-TECH AMERICA, INC.
Supplier Number: AN0-8851
Frequently Bought Together
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Supplier: MilliporeSigma
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Leakbuster™ Specimen Containers
Supplier: Starplex
Optical Lens Cleaner, FoxWest Sales
Supplier: Decon Labs
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Alkaline Batteries
Supplier: DANTONA INDUSTRIES INC SE
Difco™ Agar Soldifying Agent, BD Diagnostics
Supplier: BD
Gibson Assembly Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
Potassium Sodium Tartrate
Supplier: Ward's Science
Pierce™ HisPur™ Cobalt Superflow Agarose, Thermo Scientific
Supplier: Thermo Scientific
Orion™ Fluoride Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Iodomethane ≥99.5% stabilized
Supplier: Thermo Scientific Chemicals
PIPET CAL M12 ISO8655 10R/3V AL
Supplier: COVARIANCE SV
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Decalcifier, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International