AMINO ACID ANALYZER PF4 BUFFER
Catalog # 76522-114
Supplier: HITACHI HIGH-TECH AMERICA, INC.
AMINO ACID ANALYZER PF4 BUFFER
Catalog # 76522-114
Supplier: HITACHI HIGH-TECH AMERICA, INC.
Supplier Number: AN0-5034
Frequently Bought Together
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Supplier: MilliporeSigma
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Puritan® Standard Polyester Tipped Applicator, Polystyrene Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
Sulfuric acid 95-98% ACS, FCC, VWR Chemicals BDH®
Supplier: BDH
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Supplier: Starplex
VWR® Blood Bank Saline Solutions
Supplier: VWR International
Optical Lens Cleaner, FoxWest Sales
Supplier: Decon Labs
Solvent Resistant Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Prinzing® Ball Valve Lockouts, Brady Worldwide®
Supplier: Brady Worldwide
Potassium Phosphate Monobasic
Supplier: Ward's Science
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Lithium hydroxide monohydrate ≥98% ACS
Supplier: Thermo Scientific Chemicals
Penicillin : Streptomycin solution 100X, Corning®
Supplier: Corning
First Aid Central CSA First Aid Kits, Acme United
Supplier: ACME UNITED LIMITED CA