SEAL SYRINGE VALVE F/AUTOSAMPLER
Catalog # 76522-068
Supplier: HITACHI HIGH-TECH AMERICA, INC.
SEAL SYRINGE VALVE F/AUTOSAMPLER
Catalog # 76522-068
Supplier: HITACHI HIGH-TECH AMERICA, INC.
Supplier Number: 890-3234
Frequently Bought Together
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Blood Bank Saline Solutions
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
Accessories for perfectION™ Combined Ion Selective Electrode electrodes
Supplier: Mettler Toledo
AGAR BRAIN HEART INFUSION 450 G
Supplier: BECTON DICKINSON DE MEXICO, S. MX
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
SYRINGE WITH NEEDLE 5ML
Supplier: AMX INDUSTRIAL TRADE MX
Sodium cyanotrihydroborate ≥95%
Supplier: Thermo Scientific Chemicals