Disintegration Baskets, Agilent Technologies
Basket, 6-tube, 10-mesh, 1905 µm (USP)
Catalog # CAAG37-3001
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
: 37-3001
Disintegration Baskets, Agilent Technologies
Catalog # CAAG37-3001
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
Agilent offers disintegration baskets for the 100 automated disintegration apparatus.
Specifications
- Description:Basket, 6-tube, 10-mesh, 1905 µm (USP)