Orders LinkContactUsLinkComponent
CrossLab Sample Preparation, Agilent Technologies
TOP well plate tubes, 300 mg, 6 ml
  CAAG12102300
CrossLab Sample Preparation, Agilent Technologies
  CAAG12102300
CrossLab Sample Preparation, Agilent Technologies
CrossLab Sample Preparation, Agilent Technologies
TOP well plate tubes, 300 mg, 6 ml
$445.00
Pack of 30
Agilent offers a range of Sample Preparation products to support your workflows.
  • Achieve better results from your analyses, while also extending the lifetime of your column and instrument components
Our suite of sample preparation products can help you to achieve better results from your analyses, while also extending the lifetime of your column and instrument components. Our purification and filtration products provide reliable results for repeated analyses. Our QuEChERS, solid phase extraction (SPE), solid phase microextraction (SPME), and supported liquid extraction (SLE) products ensure your samples are free of interferences, even with complex matrices.
  • Description:
    TOP well plate tubes, 300 mg, 6 ml