Order Entry
Canada
Orders LinkContactUsLinkComponent
qScript® Ultra Flex Kits, Quantabio

qScript® Ultra Flex Kits, Quantabio

qScript® Ultra flex kit, 100
Catalog # CA76518-314
Supplier:   Quantabio
Supplier #:  95215-100
qScript® Ultra Flex Kits, Quantabio
Catalog # CA76518-314
Supplier:  Quantabio
CAS Number:  
qScript® Ultra Flex Kits, Quantabio
qScript® Ultra Flex Kits, Quantabio

qScript® Ultra flex kit, 100

CAS Number:  
Price
$1,076.69
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Faster, longer, tougher next gen cDNA synthesis with flexible priming.

  • Enhanced thermostability – Engineered for RT activity up to 65 °C, overcoming challenging secondary structures
  • Superior speed and ease of use – 10 minute reaction time with fewer pipetting steps
  • Maximum yield and sensitivity – Improved cDNA yield with limiting or compromised samples
  • Ultimate inhibitor resistance – Overcome a wide array of PCR inhibitors (salt, heparin, hematin, etc.)
  • Longer transcripts – Ability to reverse transcribe single targets up to 20 kb

qScript Ultra Flex Kit is easy-to-use and highly efficient for the synthesis of first-strand cDNA with flexible priming methods including Oligo-dT, Random Hexamer and Gene Specific Priming to reverse transcribe RNA to cDNA. A key component is a novel, state-of-the-art, RNase H deficient reverse transcriptase that was engineered for improved thermostability, velocity, processivity, and resistance to many common reaction inhibitors.

qScript Ultra Flex Kit contains all required components for first-strand cDNA synthesis except RNA template and gene specific primers. The cDNA product is directly compatible with downstream 2-step RT-qPCR or RT-PCR procedures.

qScript Ultra Flex Kit has an optimal reaction temperature of 55 °C and a 10-min reaction time, significantly reducing time to result while working through challenging secondary RNA structures. The lower Cq values for the engineered qScript Ultra reverse transcriptase indicate more efficient cDNA synthesis at high temperatures, critical for progressing through high GC regions and regions of RNA secondary structure.

Documents

Specifications

Specifications

  • Description:
    qScript® Ultra flex kit
  • No. of reactions:
    100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.