SPEC Disk SPE, Agilent Technologies
Bond Elut cartridge, Cartridge, SPEC C18, 3 ml, 15 mg
CAA5320320
Supplier #: A5320320
SPEC disk SPE delivers superior flow characteristics and trouble-free automation.
- No loose sorbent means no channeling and better reproducibility
- Uniform flow and extraction properties offer robust performance
- Low elution volume affords excellent concentration of the analyte, improving sensitivity
SPEC disk SPE is a product providing faster sample processing, lower costs, and high-quality results. SPEC is manufactured with ultraclean polypropylene plastic and contains a monolithic sorbent bed with bonded silica infused on a glass fiber disk. For some applications, the elution volume is very small, eliminating the dry down and reconstitution steps.
Integrated filtration protects against clogging. SPEC is available in cartridge, 96-well plate, and disk formats in a wide range of chemistries for many applications.
- Column Format:Cartridge
- Capacity:3 ml
- Description:Bond Elut cartridge
- Bed weight:15 mg
- Phase:SPEC C18