Funnel, Sartorius
Funnel with clamp, 500 ml
Catalog # CA76518-000
Supplier: Sartorius
Supplier #: 6981002
Jump to:
Product Details & Documents
Stainless steel, autoclavable.
To use with filter holders for colony counting.
Ordering information: Supplied with funnel and clamp (lid and seal must be ordered separately).
Specifications
- Capacity:500 ml
- Description:Funnel with clamp