Orders LinkContactUsLinkComponent
Interface Cone Supplies for ICP-MS
Interface Cone Supplies for ICP-MS
Sampler cone gasket, graphite, for Agilent 7700, 7800, 7850, 7900, 8800 and 8900 ICP-MS
Catalog # CAG3280-67009
Supplier #:  G3280-67009
Interface Cone Supplies for ICP-MS
Catalog # CAG3280-67009
Interface Cone Supplies for ICP-MS
Interface Cone Supplies for ICP-MS
Sampler cone gasket, graphite, for Agilent 7700, 7800, 7850, 7900, 8800 and 8900 ICP-MS
Price
$110.00
Log in for availability
Pack of 3
Product Details & Documents
Accessories and supplies for skimmer and sampling cones for use with our ICP-MS portfolio.
  • Graphite gasket for sampling cone that seals the sampling cone to the vacuum chamber interface
  • Available interface wrenches (in order to remove and install the skimmer cone) and sampling cone wrenches (used to remove the sampling cone retaining ring)
  • Cone cleaning and maintenance supplies including fixtures, cone magnifier, cotton swabs, Citranox and Alumina powder
Agilent's high quality ICP-MS systems contain interfaces that have a step-down vacuum stage, located between a pair of conical metal plates, known as interface cones. In common terminology, the first and second cones are referred to as the sample cone and skimmer cone, respectively. Agilent has all your interface cone maintenance needs covered with our range of supplies and accessories.
A range of accessories, components and spare parts are available including cone tools, magnifier and cleaning fixtures, sampling cone carbon/graphite gaskets, cotton cleaning swabs, Citranox and Alumina powder.
Specifications
  • Description:
    Sampler cone gasket, graphite, for Agilent 7700, 7800, 7850, 7900, 8800 and 8900 ICP-MS