Order Entry
Canada
Orders LinkContactUsLinkComponent
 

CrossLab Split Liners for GC, Agilent Technologies

Inlet liner, split, focus, glass wool, deactivated, 6.25 mm, 935 µl, 4×78.5 mm
Catalog # CA76514-580
 :  210-4004-5
CrossLab Split Liners for GC, Agilent Technologies
Catalog # CA76514-580
CrossLab Split Liners for GC, Agilent Technologies
CrossLab Split Liners for GC, Agilent Technologies

Inlet liner, split, focus, glass wool, deactivated, 6.25 mm, 935 µl, 4×78.5 mm

Price
224,00 $
Log in for availability
Pack of 5
Product Details & Documents

Agilent split liners are made to strict dimensional specifications for optimal inlet performance.

  • Feature Agilent’s exclusive deactivation;Agilent recommends part number 5183-4647 as the top split liner choice

Split injections are intended for use with a split/splitless injection port in gas chromatography analysis. Split liners tend to have a slightly smaller outer diameter to allow for gas flow. For ease of use and reproducibility, the liners have a positioning bead, a restriction to secure the position of the glass wool, and a feature to consistently self-position to the recommended height.;Agilent split liners are made to strict dimension specifications for optimal inlet performance and feature the tightest tolerances for od, id, taper and glass wool placement.

Specifications
  • Ext.Ø:
    6.25 mm
  • Volume:
    935 µl
  • Description:
    Inlet liner, split, focus, glass wool, deactivated
  • I.D.×L:
    4×78.5 mm