CrossLab Sample Preparation, Agilent Technologies
Reservoir, 140 ml
Catalog # CA12131042
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Supplier #: 12131042
CrossLab Sample Preparation, Agilent Technologies
Catalog # CA12131042
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
Agilent offers a range of Sample Preparation products to support your workflows.
- Achieve better results from your analyses, while also extending the lifetime of your column and instrument components
Our suite of sample preparation products can help you to achieve better results from your analyses, while also extending the lifetime of your column and instrument components. Our purification and filtration products provide reliable results for repeated analyses. Our QuEChERS, solid phase extraction (SPE), solid phase microextraction (SPME), and supported liquid extraction (SLE) products ensure your samples are free of interferences, even with complex matrices.
Specifications
- Description:Reservoir, 140 ml