Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Lab Pack Poly Drums, Eagle Mfg

Lab pack open head plastic barrel drum with 1×2 and 1x¾" bung holes, metal lever-lock, 16 gauge metal lever band, 30 gal., 18 lbs., 19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height", Black, Bung, 1H2/X120/S, 1H2/Y180/S
  76502-664
 :  1601MBLKBG
Lab Pack Poly Drums, Eagle Mfg
  76502-664
Lab Pack Poly Drums, Eagle Mfg
Lab Pack Poly Drums, Eagle Mfg

Lab pack open head plastic barrel drum with 1×2 and 1x¾" bung holes, metal lever-lock, 16 gauge metal lever band, 30 gal., 18 lbs., 19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height", Black, Bung, 1H2/X120/S, 1H2/Y180/S

 
$135.69
   
Each
 
 

These lab pack drums are constructed of blow-molded HDPE and are durable and lightweight.

  • Chemical and weather resistant
  • UV protection
  • Approved for use with packing groups i, ii and iii

Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration. For use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials.

 : UN certified. UL rating. Food grade quality - FDA 21 CFR 177.1520.

 
  • Capacity:
    30 gal.
  • Color:
    Black
  • Description:
    Lab pack open head plastic barrel drum with 1×2 and 1x¾" bung holes, metal lever-lock, 16 gauge metal lever band
  • Dimensions:
    19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height"
  • UN/DOT Rating:
    1H2/X120/S, 1H2/Y180/S
  • Weight:
    18 lbs.
  • Lid type:
    Bung