Lab Pack Poly Drums, Eagle Mfg
Lab pack open head plastic barrel drum with 1×2 and 1x¾" bung holes, metal lever-lock, 30 gal., 18 lbs., 19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height", Blue, Bung, 1H2/X120/S, 1H2/Y180/S
Catalog # 76502-662
Supplier: Eagle Manufacturing
Supplier #: 1601MBBG
Jump to:
Product Details & Documents
These lab pack drums are constructed of blow-molded HDPE and are durable and lightweight.
- Chemical and weather resistant
- UV protection
- Approved for use with packing groups i, ii and iii
Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration. For use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials.
Certifications: UN certified. UL rating. Food grade quality - FDA 21 CFR 177.1520.
Documents
Eagle MFG Poly Drums
Product Literature
Specifications
- Capacity:30 gal.
- Color:Blue
- Description:Lab pack open head plastic barrel drum with 1×2 and 1x¾" bung holes, metal lever-lock
- Dimensions:19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height"
- UN/DOT Rating:1H2/X120/S, 1H2/Y180/S
- Weight:18 lbs.
- Lid type:Bung
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



