Lab Pack Poly Drums, Eagle Mfg
Lab pack open head plastic barrel drum with 1×2 and 1x¾" bung holes, plastic lever-lock, 30 gal., 18 lbs., 19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height", Black, Bung, 1H2/X100/S, 1H2/Y180/S
Catalog # 76502-660
Supplier: Eagle Manufacturing
: 1601BLKBG
Jump to:
Product Details & Documents
These lab pack drums are constructed of blow-molded HDPE and are durable and lightweight.
- Chemical and weather resistant
- UV protection
- Approved for use with packing groups i, ii and iii
Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration. For use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials.
Certifications: UN certified. UL rating. Food grade quality - FDA 21 CFR 177.1520.
Documents
Eagle MFG Poly Drums
Product Literature
Specifications
- Capacity:30 gal.
- Color:Black
- Description:Lab pack open head plastic barrel drum with 1×2 and 1x¾" bung holes, plastic lever-lock
- Dimensions:19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height"
- UN/DOT Rating:1H2/X100/S, 1H2/Y180/S
- Weight:18 lbs.
- Lid type:Bung



