Order Entry
Canada
Orders LinkContactUsLinkComponent
Lab Pack Poly Drums, Eagle Mfg

Lab Pack Poly Drums, Eagle Mfg

Lab pack open head plastic barrel drum, plastic lever-lock, 30 gal., 16 lbs., 19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height, Red, Regular, 1H2/X100/S, 1H2/Y180/S
Catalog # 76502-654
Supplier:   Eagle Manufacturing
Supplier #:  1601RED
Lab Pack Poly Drums, Eagle Mfg
Catalog # 76502-654
Lab Pack Poly Drums, Eagle Mfg
Lab Pack Poly Drums, Eagle Mfg

Lab pack open head plastic barrel drum, plastic lever-lock, 30 gal., 16 lbs., 19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height, Red, Regular, 1H2/X100/S, 1H2/Y180/S

Price
$140.12
Log in for availability
Each
Product Details & Documents

These lab pack drums are constructed of blow-molded HDPE and are durable and lightweight.

  • Chemical and weather resistant
  • UV protection
  • Approved for use with packing groups i, ii and iii

Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration. For use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials.

Certifications: UN certified. UL rating. Food grade quality - FDA 21 CFR 177.1520.

Documents

Specifications
  • Capacity:
    30 gal.
  • Color:
    Red
  • Description:
    Lab pack open head plastic barrel drum, plastic lever-lock
  • Dimensions:
    19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height
  • UN/DOT Rating:
    1H2/X100/S, 1H2/Y180/S
  • Weight:
    16 lbs.
  • Lid type:
    Regular
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.