KIT LOW DISPERSION PIPNG CHROMASTER 5280
Catalog # 76501-088
Supplier: HITACHI HIGH-TECH AMERICA, INC.
KIT LOW DISPERSION PIPNG CHROMASTER 5280
Catalog # 76501-088
Supplier: HITACHI HIGH-TECH AMERICA, INC.
Supplier Number: 892-0826
Frequently Bought Together
MBP DNA AWAY™
Supplier: Thermo Scientific
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Ethyl Alcohol Denatured
Supplier: Ward's Science
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Phenols, CHEMets Visual Kit, CHEMetrics
Supplier: CHEMetrics
Phenolphthalein 1% (w/v) in 95% (v/v) alcohol indicator, neutralized
Supplier: Ricca Chemical
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
VWR® Disposable Petri Dishes
Supplier: VWR International
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
VWR® Reagent alcohol
Supplier: VWR International
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
SYRINGE WITH NEEDLE 5ML
Supplier: AMX INDUSTRIAL TRADE MX
Orion™ Star™ A325 pH/Conductivity Portable Multiparameter Meter, Thermo Scientific
Supplier: Thermo Fisher Scientific
Prepared Blood Agar Media
Supplier: Culture Media & Supplies